3-Chloro-4-hydroxybenzoic acid, (max. 6% H₂O) ≥97%

Waarschuwing

A12134.09EA 28.1 EUR
A12134.09
3-Chloro-4-hydroxybenzoic acid, (max. 6% H₂O) ≥97%
3-Chloro-4-hydroxybenzoic acid
Formule: C₇H₅ClO₃
MW: 172,57 g/mol
Kookpunt: 331 °C (1013 hPa)
Smeltpunt: 168…170 °C
MDL-nummer: MFCD00002549
CAS-nummer: 3964-58-7
EINECS: 223-574-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR