ALFA88697
88697.03EA 21.7 EUR
ALFA88697.03 ALFA88697.09
ALFA88697
For reducing organic nitro compounds, in the manufacture of sulfur dyes, in ore flotation, and in the preparation of metal sulfide finishes
Bestel nu