L(+)-Aspartic acid
EDQMA1330000EA
234
EUR
EDQMA1330000
L(+)-Aspartic acid
L(+)-Aspartic acid
Formule:
C₄H₇NO₄ MW: 133,1 g/mol Kookpunt: 311 °C (1013 hPa) Smeltpunt: 251 °C Dichtheid: 1,6613 g/cm³ (13 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00002616 CAS-nummer: 56-84-8 EINECS: 200-291-6 Merck Index: 13,00845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...