Barbituric acid

EDQMY0000762EA 234 EUR
EDQMY0000762
Barbituric acid
Barbituric acid
Formule: C₄H₄N₂O₃
MW: 128,09 g/mol
Kookpunt: 441 °C (1013 hPa)
Smeltpunt: 250…252 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006666
CAS-nummer: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR