(2,4-Dichlorophenyl)acetic acid
Leverancier: EHRENSTORFER GMBH
|
Waarschuwing
|
EHERC12468000EA
44.7
EUR
EHERC12468000
(2,4-Dichlorophenyl)acetic acid
(2,4-Dichlorophenyl)acetic acid
Formule:
(C₆H₃Cl₂)CH₂COOH MW: 205,04 g/mol Kookpunt: 316 °C (1013 hPa) Smeltpunt: 132…133 °C |
MDL-nummer:
MFCD00004318 CAS-nummer: 19719-28-9 EINECS: 243-248-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...