Diisobutylamine 2 mol/l
Leverancier: Bernd Kraft
KRAF20406.4700EA
418
EUR
KRAF20406.4700
KRAF20406.3700
KRAF20406.2700
Diisobutylamine 2 mol/l
Diisobutylamine
Formule:
C₈H₁₉N MW: 129,25 g/mol |
MDL-nummer:
MFCD00008930 CAS-nummer: 110-96-3 EINECS: 203-819-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...