4-Ethylmorpholine ≥99.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
Gevaar
TCIAE0145-500MLEA 41.8 EUR
TCIAE0145-500ML TCIAE0145-25ML
4-Ethylmorpholine ≥99.0% (by GC, titration analysis)
4-Ethylmorpholine
Formule: C₆H₁₃NO
MW: 115,18 g/mol
Kookpunt: 139°C
Smeltpunt: –63 °C
Vlampunt: 36°C
MDL-nummer: MFCD00006177
CAS-nummer: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8,II
https://be.vwr.com/stibo/web/std.lang.all/77/26/28967726.jpg
Bestel nu
Specificaties testresultaten
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %