Quinoxaline ≥99.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAQ0020-25GEA 57.2 EUR
TCIAQ0020-25G
Quinoxaline ≥99.0%
Quinoxaline
Formule: C₈H₆N₂
MW: 130,15 g/mol
Kookpunt: 226°C
Smeltpunt: 30…32 °C
MDL-nummer: MFCD00006719
CAS-nummer: 91-19-0
EINECS: 202-047-4
Merck Index: 12,08263
https://be.vwr.com/stibo/web/std.lang.all/36/86/28993686.jpg

Bestel nu

Specificaties testresultaten

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR