2-Ethylfuran ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAE0454-10MLEA 49.5 EUR
TCIAE0454-10ML
2-Ethylfuran ≥98.0%
2-Ethylfuran
Formule: C₆H₈O
MW: 96,13 g/mol
Kookpunt: 92°C
Vlampunt: -2°C
MDL-nummer: MFCD00003259
CAS-nummer: 3208-16-0
EINECS: 221-714-0
UN: 1993
ADR: 3,II
https://be.vwr.com/stibo/web/std.lang.all/79/69/28967969.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR