Methyldiphenylsilane ≥95.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAD1825-25MLEA 81.4 EUR
TCIAD1825-25ML
Methyldiphenylsilane ≥95.0%
Methyldiphenylsilane
Formule: C₁₃H₁₄Si
MW: 198,34 g/mol
Kookpunt: 91°C
Vlampunt: 120°C
MDL-nummer: MFCD00008255
CAS-nummer: 776-76-1
EINECS: 212-281-9
https://be.vwr.com/stibo/web/std.lang.all/86/10/28978610.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR