α-Bromo-2,3,4,5-tetrafluorotoluene ≥97.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAT1885-5GEA
161
EUR
TCIAT1885-5G
α-Bromo-2,3,4,5-tetrafluorotoluene ≥97.0% (by GC, titration analysis)
α-Bromo-2,3,4,5-tetrafluorotoluene
Formule:
C₇H₃BrF₄ MW: 243 g/mol Kookpunt: 110°C |
MDL-nummer:
MFCD01631453 CAS-nummer: 53001-71-1 UN: 3265 ADR: 8,III |
Specificaties testresultaten
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...