2,6-Dibromo-4-fluoroaniline ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAD2570-25GEA 313 EUR
TCIAD2570-25G
2,6-Dibromo-4-fluoroaniline ≥98.0%
2,6-Dibromo-4-fluoroaniline
Formule: C₆H₄Br₂FN
MW: 268,91 g/mol
Smeltpunt: 68 °C
MDL-nummer: MFCD00041445
CAS-nummer: 344-18-3
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/95/43/28979543.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Amber to Dark purple powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR