Dichlooracetonitril ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAD2510-25GEA 60.5 EUR
TCIAD2510-25G TCIAD2510-500G
Dichlooracetonitril ≥98.0%
Dichlooracetonitril
Formule: C₂HCl₂N
MW: 109,94 g/mol
Kookpunt: 112°C
Vlampunt: 35°C
MDL-nummer: MFCD00040886
CAS-nummer: 3018-12-0
EINECS: 221-159-4
UN: 2920
ADR: 8,II
https://be.vwr.com/stibo/web/std.lang.all/94/46/28979446.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR