6-Methylcoumarin ≥99.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAM0189-25GEA 17.6 EUR
TCIAM0189-25G
6-Methylcoumarin ≥99.0%
6-Methylcoumarin
Formule: C₁₀H₈O₂
MW: 160,17 g/mol
Kookpunt: 200°C
Smeltpunt: 72…74 °C
MDL-nummer: MFCD00006875
CAS-nummer: 92-48-8
EINECS: 202-158-8
https://be.vwr.com/stibo/web/std.lang.all/35/27/28973527.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR