4-Methoxybenzophenone ≥98.0%

Leverancier: TCI EUROPE

TCIAM0357-25GEA 40.7 EUR
TCIAM0357-25G
4-Methoxybenzophenone ≥98.0%
4-Methoxybenzophenone
Formule: C₁₄H₁₂O₂
MW: 212,25 g/mol
Kookpunt: 193°C
Smeltpunt: 59…62 °C
MDL-nummer: MFCD00008403
CAS-nummer: 611-94-9
EINECS: 210-285-5
https://be.vwr.com/stibo/web/std.lang.all/37/76/28973776.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 63.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR