(±)-2-Aminooctane ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAM0337-1GEA 72.6 EUR
TCIAM0337-1G
(±)-2-Aminooctane ≥98.0%
(±)-2-Aminooctane
Formule: C₈H₁₉N
MW: 129,25 g/mol
Kookpunt: 165°C
Vlampunt: 50°C
MDL-nummer: MFCD00008103
CAS-nummer: 693-16-3
EINECS: 211-744-2
UN: 2734
ADR: 8,II

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR