4-Methyl-o-fenyleendiamine ≥98.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAD0125-25GEA
70.4
EUR
TCIAD0125-25G
4-Methyl-o-fenyleendiamine ≥98.0%
4-Methyl-o-fenyleendiamine
Formule:
C₇H₁₀N₂ MW: 122,17 g/mol Kookpunt: 265°C Smeltpunt: 86…89 °C |
MDL-nummer:
MFCD00007728 CAS-nummer: 496-72-0 EINECS: 207-826-2 |
Specificaties testresultaten
Appearance | White to Gray to Brown powder to lump |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 89.0 to 92.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...