3-Methoxy-2-nitropyridine ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAM1359-25GEA 135 EUR
TCIAM1359-25G
3-Methoxy-2-nitropyridine ≥98.0%
3-Methoxy-2-nitropyridine
Formule: C₆H₆N₂O₃
MW: 154,13 g/mol
MDL-nummer: MFCD00222276
CAS-nummer: 20265-37-6
https://be.vwr.com/stibo/web/std.lang.all/53/71/28975371.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR