O-Cymene ≥99.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAC0799-0.1MLEA
94.6
EUR
TCIAC0799-0.1ML
O-Cymene ≥99.0%
O-Cymene
Formule:
C₁₀H₁₄ MW: 134,22 g/mol Smeltpunt: –67,9 °C Vlampunt: 50°C |
MDL-nummer:
MFCD00008888 CAS-nummer: 527-84-4 EINECS: 208-426-0 UN: 2046 ADR: 3,III |
Specificaties testresultaten
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...