m-Terphenyl ≥98.0% (by GC)

Leverancier: TCI EUROPE
Waarschuwing

TCIAT0018-500GEA 311 EUR
TCIAT0018-500G TCIAT0018-25G TCIAT0018-100G
m-Terphenyl ≥98.0% (by GC)
m-Terphenyl
Formule: C₁₈H₁₄
MW: 230,31 g/mol
Kookpunt: 365°C
Smeltpunt: 86…87 °C
MDL-nummer: MFCD00003059
CAS-nummer: 92-06-8
EINECS: 202-122-1
UN: 3077
ADR: 9,III
https://be.vwr.com/stibo/web/std.lang.all/43/60/28994360.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 90.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR