Triphenylarsine ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAT0508-5GEA 103 EUR
TCIAT0508-5G
Triphenylarsine ≥98.0%
Triphenylarsine
Formule: C₁₈H₁₅As
MW: 306,24 g/mol
Kookpunt: 175°C
Smeltpunt: 58 °C
MDL-nummer: MFCD00002994
CAS-nummer: 603-32-7
EINECS: 210-032-9
UN: 3465
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/47/49/28994749.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 60.0 to 62.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR