2,5-Xylylacetic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAD3779-25GEA
23.1
EUR
TCIAD3779-25G
2,5-Xylylacetic acid ≥98.0% (by GC, titration analysis)
2,5-Xylylacetic acid
Formule:
(C₆H₃(CH₃)₂)CH₂COOH MW: 164,2 g/mol |
MDL-nummer:
MFCD00014369 CAS-nummer: 13612-34-5 EINECS: 237-097-6 |
Specificaties testresultaten
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...