(3,4-Dimethoxyphenyl)acetic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI EUROPE
Waarschuwing

TCIAD0640-25GEA 26.4 EUR
TCIAD0640-25G
(3,4-Dimethoxyphenyl)acetic acid ≥98.0% (by titrimetric analysis)
(3,4-Dimethoxyphenyl)acetic acid
Formule: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196,2 g/mol
Smeltpunt: 97…99 °C
MDL-nummer: MFCD00004335
CAS-nummer: 93-40-3
EINECS: 202-244-5
https://be.vwr.com/stibo/web/std.lang.all/55/16/28975516.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 101.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR