3',4'-Dichloroacetophenone ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAD0311-25GEA 178 EUR
TCIAD0311-25G
3',4'-Dichloroacetophenone ≥98.0%
3',4'-Dichloroacetophenone
Formule: C₈H₆Cl₂O
MW: 189,04 g/mol
Kookpunt: 121°C
Smeltpunt: 44…46 °C
MDL-nummer: MFCD00000553
CAS-nummer: 2642-63-9
EINECS: 220-146-0
https://be.vwr.com/stibo/web/std.lang.all/47/75/28974775.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR