2,6-Dichlorophenyl isocyanate ≥98.0%
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAD3668-5GEA
61.6
EUR
TCIAD3668-5G
2,6-Dichlorophenyl isocyanate ≥98.0%
2,6-Dichlorophenyl isocyanate
Formule:
C₇H₃Cl₂NO MW: 188,01 g/mol Kookpunt: 130°C Smeltpunt: 42…44 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00002003 CAS-nummer: 39920-37-1 EINECS: 254-699-4 UN: 2250 ADR: 6.1,II |
Specificaties testresultaten
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...