Dimethyl azelate ≥98.0% (by GC)

Leverancier: TCI EUROPE

TCIAA1308-25MLEA 81.4 EUR
TCIAA1308-25ML
Dimethyl azelate ≥98.0% (by GC)
Dimethyl azelate
Formule: C₁₁H₂₀O₄
MW: 216,28 g/mol
Kookpunt: 140°C
MDL-nummer: MFCD00025898
CAS-nummer: 1732-10-1
EINECS: 217-060-0
https://be.vwr.com/stibo/web/std.lang.all/92/08/28929208.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR