2-Chlorobenzylamine ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAC1065-25GEA 39.6 EUR
TCIAC1065-25G
2-Chlorobenzylamine ≥98.0%
2-Chlorobenzylamine
Formule: C₇H₈ClN
MW: 141,6 g/mol
Kookpunt: 219°C
Vlampunt: 88°C
MDL-nummer: MFCD00008108
CAS-nummer: 89-97-4
EINECS: 201-955-8
UN: 2735
ADR: 8,III
https://be.vwr.com/stibo/web/std.lang.all/83/28/28938328.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR