4-Chlorobenzylamine ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAC1090-25GEA 66 EUR
TCIAC1090-25G
4-Chlorobenzylamine ≥98.0%
4-Chlorobenzylamine
Formule: C₇H₈ClN
MW: 141,6 g/mol
Kookpunt: 214°C
Vlampunt: 90°C
MDL-nummer: MFCD00008121
CAS-nummer: 104-86-9
EINECS: 203-245-3
UN: 2735
ADR: 8,III
https://be.vwr.com/stibo/web/std.lang.all/83/53/28938353.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR