4-Methylbenzylamine ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAM0864-25MLEA 110 EUR
TCIAM0864-25ML
4-Methylbenzylamine ≥98.0%
4-Methylbenzylamine
Formule: C₈H₁₁N
MW: 121,18 g/mol
Kookpunt: 196°C
Smeltpunt: 20…21 °C
Vlampunt: 80°C
MDL-nummer: MFCD00008123
CAS-nummer: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III
https://be.vwr.com/stibo/web/std.lang.all/43/63/28974363.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR