6-Nitro-o-toluidine ≥99.0%
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAN0467-25GEA
126
EUR
TCIAN0467-25G
6-Nitro-o-toluidine ≥99.0%
6-Nitro-o-toluidine
Formule:
C₇H₈N₂O₂ MW: 152,15 g/mol Kookpunt: 124°C Smeltpunt: 92…94 °C |
MDL-nummer:
MFCD00007744 CAS-nummer: 570-24-1 EINECS: 209-329-6 UN: 2660 ADR: 6.1,III |
Specificaties testresultaten
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 93.0 to 97.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...