4-(Difluoromethoxy)aniline ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAD3457-5GEA 84.7 EUR
TCIAD3457-5G
4-(Difluoromethoxy)aniline ≥98.0%
4-(Difluoromethoxy)aniline
Formule: C₇H₇F₂NO
MW: 159,14 g/mol
Kookpunt: 90°C
MDL-nummer: MFCD00085005
CAS-nummer: 22236-10-8
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/04/92/28980492.jpg

Bestel nu

Specificaties testresultaten

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific gravity (20/20) 1.2840 to 1.2880
Refractive index n20/D 1.5040 to 1.5080

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR