4-Bromothioanisole ≥99.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAB2024-25GEA
94.6
EUR
TCIAB2024-25G
4-Bromothioanisole ≥99.0%
4-Bromothioanisole
Formule:
C₇H₇BrS MW: 203,1 g/mol Kookpunt: 250°C Smeltpunt: 35 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00000102 CAS-nummer: 104-95-0 EINECS: 203-255-8 ADR: 9, |
Specificaties testresultaten
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 36.0 to 40.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...