3-Chloro-4-fluoroanisole ≥97.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAC2217-5GEA 58.3 EUR
TCIAC2217-5G
3-Chloro-4-fluoroanisole ≥97.0%
3-Chloro-4-fluoroanisole
Formule: C₇H₆ClFO
MW: 160,58 g/mol
Vlampunt: 79°C
MDL-nummer: MFCD00070778
CAS-nummer: 202925-07-3

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR