4-Chlorobenzophenone ≥98.0%

TCIAC1485-25GEA 17.6 EUR
TCIAC1485-25G
4-Chlorobenzophenone ≥98.0%
4-Chlorobenzophenone
Formule: C₁₃H₉ClO
MW: 216,67 g/mol
Kookpunt: 170°C
Smeltpunt: 68…70 °C
MDL-nummer: MFCD00000622
CAS-nummer: 134-85-0
EINECS: 205-160-7
https://be.vwr.com/stibo/web/std.lang.all/87/04/28938704.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR