2'-Methoxyacetophenone ≥97.0%
Leverancier: TCI EUROPE
TCIAM1445-250GEA 662 EUR
TCIAM1445-250G TCIAM1445-25G
2'-Methoxyacetophenone ≥97.0%
2'-Methoxyacetophenone
Formule: C₉H₁₀O₂
MW: 150,18 g/mol
Vlampunt: 108°C
MDL-nummer: MFCD00008725
CAS-nummer: 579-74-8
EINECS: 209-446-2
https://be.vwr.com/stibo/web/std.lang.all/56/22/28975622.jpg
Bestel nu
Specificaties testresultaten
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %