1-Ethyl-2-iodobenzene ≥97.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAE0512-5GEA 62.7 EUR
TCIAE0512-5G
1-Ethyl-2-iodobenzene ≥97.0%
1-Ethyl-2-iodobenzene
Formule: C₈H₉I
MW: 232,06 g/mol
Kookpunt: 221°C
MDL-nummer: MFCD00040871
CAS-nummer: 18282-40-1
https://be.vwr.com/stibo/web/std.lang.all/80/23/28968023.jpg

Bestel nu

Specificaties testresultaten

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR