2,3,4-Trifluorobenzoyl chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAT1868-5GEA
165
EUR
TCIAT1868-5G
2,3,4-Trifluorobenzoyl chloride ≥98.0% (by titrimetric analysis)
2,3,4-Trifluorobenzoyl chloride
Formule:
C₇H₂ClF₃O MW: 194,54 g/mol Vlampunt: 92°C |
MDL-nummer:
MFCD00075078 CAS-nummer: 157373-08-5 UN: 3265 ADR: 8,III |
Specificaties testresultaten
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...