2,6-Difluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAD2798-25GEA
70.4
EUR
TCIAD2798-25G
2,6-Difluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
2,6-Difluorobenzoyl chloride
Formule:
C₇H₃ClF₂O MW: 176,55 g/mol Vlampunt: 46°C |
MDL-nummer:
MFCD00000659 CAS-nummer: 18063-02-0 EINECS: 241-971-2 UN: 2920 ADR: 8,II |
Specificaties testresultaten
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...