2-Amino-6-chloropyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI EUROPE
Waarschuwing

TCIAA2263-5GEA 60.5 EUR
TCIAA2263-5G
2-Amino-6-chloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-chloropyridine
Formule: C₅H₅ClN₂
MW: 128,56 g/mol
MDL-nummer: MFCD00234068
CAS-nummer: 45644-21-1
https://be.vwr.com/stibo/web/std.lang.all/04/39/28930439.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Yellow to Orange powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR