2-Chloropyrimidine ≥99.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAC1050-25GEA 53.9 EUR
TCIAC1050-25G
2-Chloropyrimidine ≥99.0%
2-Chloropyrimidine
Formule: C₄H₃ClN₂
MW: 114,53 g/mol
Kookpunt: 91°C
Smeltpunt: 65…66 °C
MDL-nummer: MFCD00006060
CAS-nummer: 1722-12-9
EINECS: 217-020-2
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/83/09/28938309.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 63.0 to 67.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR