4-Pyridone ≥99.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAH0332-25GEA
80.3
EUR
TCIAH0332-25G
4-Pyridone ≥99.0%
4-Pyridone
Formule:
C₅H₅NO MW: 95,1 g/mol Smeltpunt: 146…148 °C |
MDL-nummer:
MFCD00040458 CAS-nummer: 626-64-2 EINECS: 210-958-3 |
Specificaties testresultaten
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 148.0 to 153.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...