L(-)-Mannose ≥98.0% (by HPLC)
Leverancier: TCI EUROPE
TCIAM1308-1GEA
142
EUR
TCIAM1308-1G
L(-)-Mannose ≥98.0% (by HPLC)
L(-)-Mannose
Formule:
C₆H₁₂O₆ MW: 180,16 g/mol Smeltpunt: 132 °C |
CAS-nummer:
10030-80-5 EINECS: 233-080-2 |
Specificaties testresultaten
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -13.0 to -15.0 deg (C=4, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...