Butyl stearate ≥97.0% (by GC)

Leverancier: TCI EUROPE

TCIAS0077-400GEA 35.2 EUR
TCIAS0077-400G TCIAS0077-25G
Butyl stearate ≥97.0% (by GC)
Butyl stearate
Formule: C₂₂H₄₄O₂
MW: 340,59 g/mol
Smeltpunt: 21 °C
MDL-nummer: MFCD00026669
CAS-nummer: 123-95-5
EINECS: 204-666-5
Merck Index: 13,01589
https://be.vwr.com/stibo/web/std.lang.all/39/32/28993932.jpg

Bestel nu

Specificaties testresultaten

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Freezing point 25.0 to 27.0 deg-C
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR