Octadecyltriethoxysilane ≥85.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAO0165-25GEA
97.9
EUR
TCIAO0165-25G
Octadecyltriethoxysilane ≥85.0%
Octadecyltriethoxysilane
Formule:
C₂₄H₅₂O₃Si MW: 416,76 g/mol Kookpunt: 169°C Vlampunt: 189°C |
MDL-nummer:
MFCD00039875 CAS-nummer: 7399-00-0 EINECS: 230-995-9 |
Specificaties testresultaten
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...