Dipropyl sulphite ≥99.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAD0935-25MLEA 68.2 EUR
TCIAD0935-25ML
Dipropyl sulphite ≥99.0%
Dipropyl sulphite
Formule: C₆H₁₄O₃S
MW: 166,24 g/mol
Kookpunt: 192°C
MDL-nummer: MFCD00059404
CAS-nummer: 623-98-3
ADR: 9,
https://be.vwr.com/stibo/web/std.lang.all/60/81/28976081.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR