Tetrachloropyrimidine ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAT1217-5GEA 68.2 EUR
TCIAT1217-5G
Tetrachloropyrimidine ≥98.0%
Tetrachloropyrimidine
Formule: C₄Cl₄N₂
MW: 217,87 g/mol
Smeltpunt: 70 °C
MDL-nummer: MFCD00006062
CAS-nummer: 1780-40-1
EINECS: 217-228-3
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/52/62/28995262.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR