Tetramethylammonium hydroxide pentahydrate ≥97.0%

Leverancier: TCI EUROPE
Gevaar

TCIAT1404-500GEA 735 EUR
TCIAT1404-500G TCIAT1404-100G TCIAT1404-25G
Tetramethylammonium hydroxide pentahydrate ≥97.0%
Tetramethylammonium hydroxide pentahydrate
Formule: C₄H₁₃NO·5H₂O
MW: 181,23 g/mol
Smeltpunt: 65…68 °C
MDL-nummer: MFCD00149566
CAS-nummer: 10424-65-4
UN: 3423
ADR: 8,II
https://be.vwr.com/stibo/web/std.lang.all/54/27/28995427.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Melting point 69.0 to 75.0 deg-C
Water 44.0 to 52.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR