Tetraphenyltin ≥98.0% (W)
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAT0165-25GEA
77
EUR
TCIAT0165-25G
Tetraphenyltin ≥98.0% (W)
Tetraphenyltin
Formule:
C₂₄H₂₀Sn MW: 427,13 g/mol Smeltpunt: 223 °C |
MDL-nummer:
MFCD00003006 CAS-nummer: 595-90-4 EINECS: 209-872-9 UN: 3146 ADR: 6.1,III |
Specificaties testresultaten
Appearance | White to Orange to Green powder to crystal |
Purity (Gravimetric Method) | 98.0 to 102.0 % |
Melting point | 227.0 to 231.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...