DL-O-Tyrosine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI EUROPE
Waarschuwing

TCIAT0786-1GEA 582 EUR
TCIAT0786-1G
DL-O-Tyrosine ≥98.0% (by HPLC, titration analysis)
DL-O-Tyrosine
Formule: C₉H₁₁NO₃
MW: 181,19 g/mol
MDL-nummer: MFCD00021725
CAS-nummer: 2370-61-8
EINECS: 219-134-8
https://be.vwr.com/stibo/web/std.lang.all/49/10/28994910.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in HCl (1+3) very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR