5-Nitro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAN0214-1GEA
35.2
EUR
TCIAN0214-1G
5-Nitro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
5-Nitro-1,10-phenanthroline
Formule:
C₁₂H₇N₃O₂ Smeltpunt: 203 °C |
MDL-nummer:
MFCD00004981 CAS-nummer: 4199-88-6 EINECS: 224-097-6 |
Specificaties testresultaten
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 200.0 to 203.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...